Phenyl Sepharose™ 6 Fast Flow (Low Sub) Hydrophobic interaction chromatography media
Leverandør: Cytiva
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
Sepharose™
17-0965-10EA 1186.67 DKK
17-0965-10 17-0973-05 17-0965-05 17-0973-10
Phenyl Sepharose™ 6 Fast Flow (Low Sub) Hydrophobic interaction chromatography media
Protein oprensnings Protein Purification Resins
Phenyl Sepharose™ 6 Fast Flow is a well established, standard aromatic hydrophobic interaction chromatography (HIC) medium for capture and intermediate purification requiring low to medium hydrophobicity. The hydrophilic nature of the base matrix ensures low levels of non-specific binding.
  • Standard aromatic HIC media based on Sepharose Fast Flow base matrix derivatized via uncharged, chemically-stable ether linkages
  • Designed for initial and intermediate purification requiring low to medium hydrophobicity
  • Two levels of substitution help find optimal selectivity and binding capacity for a given application
  • BioProcess medium supported for industrial applications and well-established in approved processes
  • The hydrophilic nature of the base matrix ensures low levels of non-specific binding
Phenyl Sepharose 6 Fast Flow is part of the Sepharose Fast Flow HIC platform, which has been an industrial standard for HIC chromatography during recent decades. Phenyl Sepharose 6 Fast Flow is composed of cross-linked 6% agarose beads modified with standard aromatic phenyl groups via uncharged, chemically-stable ether linkages. Two levels of ligand substitution degree help to find the optimal selectivity and binding capacity for a given application. Phenyl Sepharose 6 Fast Flow is available in a range of different bulk pack sizes and convenient pre-packed formats for easy scale-up and process development. As member of the BioProcess media range, Phenyl Sepharose 6 Fast Flow meets industrial demands with security of supply and comprehensive technical and regulatory support.
Phenyl Sepharose 6 Fast Flow is part of the Sepharose Fast Flow HIC platform, which has been an industrial standard for HIC chromatography during recent decades. Phenyl Sepharose 6 Fast Flow is composed of cross-linked 6% agarose beads modified with standard aromatic phenyl groups via uncharged, chemically-stable ether linkages. Two levels of ligand substitution degree help to find the optimal selectivity and binding capacity for a given application.