Intensifying screens, Amersham Hyperscreen™
Leverandør: Cytiva
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
RPN1662EA 2254 DKK
GEHERPN1662
Intensifying screens, Amersham Hyperscreen™
Hyperscreen™ intensifying screens are high performance calcium tungstate screens that can be used with pre-flashed film to give an increase in sensitivity while retaining a linear response that allows quantitation of results.
  • Offer rapid results and high sensitivity for detecting ³²P and ¹²⁵I in Southern, Northern, and Western blots
  • Fine grain screens give clear band resolution
  • Available in a full range of sizes