Mini portable desiccator cabinets, SICCO Mini Mobile
Leverandør: Bohlender
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
SICCO
467-0126EA 2240 DKK
467-0126 467-0129
Mini portable desiccator cabinets, SICCO Mini Mobile
Ekssikatorer Ekssikatorkabinetter
Clear polycarbonate portable desiccator with convenient handle. Door with snap fit closing and integrated gasket ensuring easy opening and tight closure. Temperature resistant from −35 to +70 °C, suitable for use under normal pressure.
  • Large opening and door with wide opening angle enable user to access the entire interior space
  • Robust construction, lightweight and portable
  • Premium model includes hygrometer and two shelves
Leveringsinformation: Supplied with desiccant which is in a handy, resealable container.