Manual tensiometer, TD 1 C
Leverandør: Lauda
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
649-0003EA 86370 DKK
649-0003
Manual tensiometer, TD 1 C
Tensiometre
Manual tensiometer for surface and interfacial tension, using a Du Noüy ring or Wilhelmy plate, and density.
Certificeringer: DIN 53914 and 53993, ASTM D971, ISO 304 and 4311.
Leveringsinformation: Supplied with two-branch Du Noüy ring, weight calibration (500 mg), glass plunger (for density), pliers, 10× glass beakers (Ø = 6 cm) and case.