2-Iodomelatonin, 98+%
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
J61498.MFEA 3470 DKK
ALFAJ61498.MF ALFAJ61498.MA
2-Iodomelatonin, 98+%
Purity: 98+%
CAS No.: 93515-00-5
Molecular Formula: C13H15IN2O2
Formula Weight: 358.18
Storage Temperature: 2°C to 8°C
Physical Form: Crystalline
MDL No.: MFCD00055216
Potent melatonin agonist