Sulfamid

Leverandør: EHRENSTORFER GMBH
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

EHERC16998170EA 588 DKK
EHERC16998170
Sulfamid
Sulfamid
Formel: H₄N₂O₂S
Molekylvægt: 96,11 g/mol
MDL Number: MFCD00011606
CAS-nummer: 7803-58-9
EINECS: 232-262-9

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR