Natriumdeoxycholat ≥97%
Leverandør: Thermo Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
J62288.14EA
384
DKK
J62288.14
J62288.30
Natriumdeoxycholat ≥97%
Natriumdeoxycholat
Formel:
C₂₄H₃₉NaO₄ Molekylvægt: 414,56 g/mol Smeltepunkt: 357…365 °C Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00064139 CAS-nummer: 302-95-4 EINECS: 206-132-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...