N-(chlormethyl)phthalimid ≥98.0% (by HPLC, titration analysis) for HPLC labeling
Leverandør: TCI EUROPE
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
TCIAA5504-5GEA
1150
DKK
TCIAA5504-5G
N-(chlormethyl)phthalimid ≥98.0% (by HPLC, titration analysis) for HPLC labeling
N-(chlormethyl)phthalimid
Formel:
C₉H₆ClNO₂ Molekylvægt: 195,6 g/mol Smeltepunkt: 123…126 °C |
MDL Number:
MFCD00005898 CAS-nummer: 17564-64-6 EINECS: 241-541-4 |
![]() |
Specifikationer Testresultater
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Argentmetric Titration) | min. 98.0 % |
Melting point | 133.0 to 137.0 deg-C |
Solubility in hot Methanol | almost transparency |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...