2-Amino-3-Brom-5-fluorpyridin 97%
Leverandør: Thermo Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Farlig
|
H37257.06EA
2520
DKK
H37257.06
2-Amino-3-Brom-5-fluorpyridin 97%
2-Amino-3-Brom-5-fluorpyridin
Formel:
C₅H₄BrFN₂ Kogepunkt: 63…65 °C Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD03092924 CAS-nummer: 869557-43-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...