2-(Trifluoromethyl)phenylboronic acid pinacol ester ≥98.0% (by GC)
Leverandør: TCI EUROPE
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
TCIAT2428-5GEA
944
DKK
TCIAT2428-5G
2-(Trifluoromethyl)phenylboronic acid pinacol ester ≥98.0% (by GC)
2-(Trifluoromethyl)phenylboronic acid pinacol ester
Formel:
C₁₃H₁₆BF₃O₂ Molekylvægt: 272,0712 g/mol Kogepunkt: 73°C |
MDL Number:
MFCD06795676 CAS-nummer: 1073339-21-5 |
Specifikationer Testresultater
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...