4-Fluoro-N-n-hexylbenzylamine, 97%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

H57865.MDEA 1240 DKK
ALFAH57865.MD ALFAH57865.03
4-Fluoro-N-n-hexylbenzylamine, 97%
Purity: 97%
CAS No.: 869944-64-9
Molecular Formula: C13H20FN
Formula Weight: 209.31
MDL No.: MFCD04482300
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR