N-Ethyl-3-fluorobenzylamine, 97%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

H56434.MDEA 495 DKK
ALFAH56434.MD ALFAH56434.03
N-Ethyl-3-fluorobenzylamine, 97%
Purity: 97%
CAS No.: 90389-85-8
Molecular Formula: C9H12FN
Formula Weight: 153.19
UN No.: 2735
MDL No.: MFCD06655514
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR