Perfluoropelargonic acid ≥95.0% (by GC, titration analysis)

Leverandør: TCI EUROPE
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

TCIAH0843-25GEA 1080 DKK
TCIAH0843-25G TCIAH0843-5G
Perfluoropelargonic acid ≥95.0% (by GC, titration analysis)
Perfluoropelargonic acid
Formel: C₉HF₁₇O₂
Molekylvægt: 464,07
MDL Number: MFCD00039605
CAS-nummer: 375-95-1
EINECS: 206-801-3
UN nummer: 1759
ADR: 8,III
https://dk.vwr.com/stibo/web/std.lang.all/08/68/28970868.jpg

Bestil nu

Specifikationer Testresultater

Appearance White to Orange to Green powder to crystaline
Purity (GC) min. 95.0 %
Purity (Neutralization titration) min. 95.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR