Perfluoropelargonic acid

Leverandør: CAMBRIDGE ISOTOPE LABS., INC.
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

CAMBULM-8066-1.2EA 0 DKK
CAMBULM-8066-1.2
Perfluoropelargonic acid
Perfluoropelargonic acid
Formel: C₉HF₁₇O₂
Molekylvægt: 464,07 g/mol
MDL Number: MFCD00039605
CAS-nummer: 375-95-1
EINECS: 206-801-3
UN nummer: 3261
ADR: 8,III

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR