Kaliumchlorid 0.075 mol/l (0.075 N)

Leverandør: Ampliqon A/S
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering


AMPQ76900.0500EA 2240 DKK
AMPQ76900.0500
Kaliumchlorid 0.075 mol/l (0.075 N)
Potassium chloride
Formel: KCl
Molekylvægt: 74,55 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00011360
CAS-nummer: 7447-40-7
EINECS: 231-211-8

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR