Methyl perfluorodecanoate ≥97.0% (by GC)
Leverandør: TCI EUROPE
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
TCIAM1916-5GEA
288
DKK
TCIAM1916-5G
Methyl perfluorodecanoate ≥97.0% (by GC)
Methyl perfluorodecanoate
Formel:
C₁₁H₃F₁₉O₂ Kogepunkt: 193°C Flammepunkt: 81°C |
MDL Number:
MFCD00153211 CAS-nummer: 307-79-9 |
![]() |
Specifikationer Testresultater
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 97.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...