3'-Acetamidoacetophenone ≥98.0% (by GC)
Leverandør: TCI EUROPE
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
TCIAA1408-25GEA
255
DKK
TCIAA1408-25G
TCIAA1408-500G
3'-Acetamidoacetophenone ≥98.0% (by GC)
3'-Acetamidoacetophenone
Formel:
C₁₀H₁₁NO₂ Molekylvægt: 177,20 g/mol |
MDL Number:
MFCD00032278 CAS-nummer: 7463-31-2 |
![]() |
Specifikationer Testresultater
Appearance | White to Light yellow to Light orange powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 127.0 to 130.0 deg-C |
Solubility in Methanol | almost transparency |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...