3-Fluoro-4-methoxybenzylamine 97%

Leverandør: Thermo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

H26607.03EA 747 DKK
H26607.03 H26607.09
3-Fluoro-4-methoxybenzylamine 97%
3-Fluoro-4-methoxybenzylamine
Formel: C₈H₁₀FNO
Molekylvægt: 155,17
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD04116360
CAS-nummer: 123652-95-9
ADR: 8,III

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR