cis-5-Octen-1-ol ≥90.0% (by GC)
Leverandør: TCI EUROPE
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
TCIAO0250-10MLEA
1560
DKK
TCIAO0250-10ML
cis-5-Octen-1-ol ≥90.0% (by GC)
cis-5-Octen-1-ol
Formel:
C₈H₁₆O Molekylvægt: 128,22 g/mol Kogepunkt: 97°C Flammepunkt: 82°C |
MDL Number:
MFCD00015569 CAS-nummer: 64275-73-6 EINECS: 264-764-9 |
Specifikationer Testresultater
Appearance | Colorless to Light yellow clear liquid |
Purity (GC) | min. 90.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...