cis-5-Octen-1-ol 95% remainder mainly trans-isomer

Leverandør: Thermo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

A19454.14EA 1260 DKK
A19454.14 A19454.06 A19454.22
cis-5-Octen-1-ol 95% remainder mainly trans-isomer
cis-5-Octen-1-ol
Formel: C₈H₁₆O
Molekylvægt: 128,22
Kogepunkt: 95 °C (25 mmHg)
Massefylde: 0,850
Flammepunkt: 82 °C (179 °F)
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00015569
CAS-nummer: 64275-73-6
EINECS: 264-764-9

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR