2-Fluoro-5-(2-methoxybenzylcarbamoyl)benzeneboronic acid, 97%
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
H54267.MDEA 367 DKK
ALFAH54267.MD ALFAH54267.03
2-Fluoro-5-(2-methoxybenzylcarbamoyl)benzeneboronic acid, 97%
Purity: 97%
Molecular Formula: C15H15BFNO4
Formula Weight: 303.10
MDL No.: MFCD20265219