3-Fluoro-5-(2-methoxybenzylcarbamoyl)benzeneboronic acid, 97%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

H54185.MDEA 988 DKK
ALFAH54185.MD ALFAH54185.03
3-Fluoro-5-(2-methoxybenzylcarbamoyl)benzeneboronic acid, 97%
Purity: 97%
Molecular Formula: C15H15BFNO4
Formula Weight: 303.10
MDL No.: MFCD20265202
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR