3-Fluoro-5-(4-fluorobenzylcarbamoyl)benzeneboronic acid, 97%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

H54182.MDEA 988 DKK
ALFAH54182.MD ALFAH54182.03
3-Fluoro-5-(4-fluorobenzylcarbamoyl)benzeneboronic acid, 97%
Purity: 97%
Molecular Formula: C14H12BF2NO3
Formula Weight: 291.06
MDL No.: MFCD20040150
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR