3-Fluoro-5-(3-fluorobenzylcarbamoyl)benzeneboronic acid, 97%
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
H54592.03EA 2050 DKK
ALFAH54592.03 ALFAH54592.MD
3-Fluoro-5-(3-fluorobenzylcarbamoyl)benzeneboronic acid, 97%
Purity: 97%
Molecular Formula: C14H12BF2NO3
Formula Weight: 291.06
MDL No.: MFCD20040226