3-Fluoro-5-(2-fluoro-5-methylbenzylcarbamoyl)benzeneboronic acid, 97%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

H54288.MDEA 495 DKK
ALFAH54288.MD ALFAH54288.03
3-Fluoro-5-(2-fluoro-5-methylbenzylcarbamoyl)benzeneboronic acid, 97%
Purity: 97%
Molecular Formula: C15H14BF2NO3
Formula Weight: 305.09
MDL No.: MFCD20091229
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR