3-Fluoro-4-(4-fluorobenzylcarbamoyl)benzeneboronic acid, 97%
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
H54563.03EA 3960 DKK
ALFAH54563.03 ALFAH54563.MD
3-Fluoro-4-(4-fluorobenzylcarbamoyl)benzeneboronic acid, 97%
Purity: 97%
Molecular Formula: C14H12BF2NO3
Formula Weight: 291.06
MDL No.: MFCD20040223