4-Chloro-3-(4-fluorobenzylcarbamoyl)benzeneboronic acid, 97%
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
H54803.03EA 2200 DKK
ALFAH54803.03 ALFAH54803.MD
4-Chloro-3-(4-fluorobenzylcarbamoyl)benzeneboronic acid, 97%
Purity: 97%
Molecular Formula: C14H12BClFNO3
Formula Weight: 307.51
MDL No.: MFCD20040250