4-Benzyl(tert-butyl)carbamoyl-2-chlorobenzeneboronic acid, 97%
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
H54514.MDEA 839 DKK
ALFAH54514.MD ALFAH54514.03
4-Benzyl(tert-butyl)carbamoyl-2-chlorobenzeneboronic acid, 97%
Purity: 97%
Molecular Formula: C18H21BClNO3
Formula Weight: 345.63
MDL No.: MFCD22125074