3-(4-Methoxybenzyloxy)-5-(trifluoromethyl)benzeneboronic acid, 98%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

H52856.03EA 2990 DKK
ALFAH52856.03 ALFAH52856.MD
3-(4-Methoxybenzyloxy)-5-(trifluoromethyl)benzeneboronic acid, 98%
Purity: 98%
CAS No.: 913835-77-5
Molecular Formula: C15H14BF3O4
Formula Weight: 326.08
MDL No.: MFCD08689490
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR