2-(Phenylsulphonyl)acetophenone ≥98.0% (by titrimetric analysis)
Leverandør: TCI EUROPE
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
TCIAP0828-5GEA
764
DKK
TCIAP0828-5G
TCIAP0828-25G
2-(Phenylsulphonyl)acetophenone ≥98.0% (by titrimetric analysis)
2-(Phenylsulphonyl)acetophenone
Formel:
C₁₄H₁₂O₃S |
MDL Number:
MFCD00025043 CAS-nummer: 3406-03-9 EINECS: 222-292-0 |
![]() |
Specifikationer Testresultater
Appearance | White to Orange to Green powder to crystal |
Purity (Neutralization titration) | min. 98.0 % |
Melting point | 91.0 to 95.0 deg-C |
Solubility in hot Toluene | very faint turbidity |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...