2-(p-Toluenesulphonyl)acetophenone ≥98.0% (by GC)
Leverandør: TCI EUROPE
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
TCIAT0908-10GEA
1380
DKK
TCIAT0908-10G
2-(p-Toluenesulphonyl)acetophenone ≥98.0% (by GC)
2-(p-Toluenesulphonyl)acetophenone
Formel:
C₁₅H₁₄O₃S |
MDL Number:
MFCD00026002 CAS-nummer: 31378-03-7 EINECS: 250-605-0 |
Specifikationer Testresultater
Appearance | White to Orange to Green powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 107.0 to 110.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...