2-Methoxy-4-(trifluoromethoxy)benzyl bromide ≥97%

Leverandør: Thermo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

H26771.03EA 1410 DKK
H26771.03 H26771.06
2-Methoxy-4-(trifluoromethoxy)benzyl bromide ≥97%
2-Methoxy-4-(trifluoromethoxy)benzyl bromide
Formel: C₉H₈BrF₃O₂
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD04115970
CAS-nummer: 886500-35-2
UN nummer: 3265
ADR: 8,II

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR