2,4-Dinitro-5-fluoroaniline ≥98.0% (by GC)

Leverandør: TCI EUROPE
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

TCIAD1637-1GEA 362 DKK
TCIAD1637-1G
2,4-Dinitro-5-fluoroaniline ≥98.0% (by GC)
2,4-Dinitro-5-fluoroaniline
Formel: C₆H₄FN₃O₄
MDL Number: MFCD00007150
CAS-nummer: 367-81-7
EINECS: 206-700-4
UN nummer: 2811
ADR: 6.1,III
https://dk.vwr.com/stibo/web/std.lang.all/84/33/28978433.jpg

Bestil nu

Specifikationer Testresultater

Appearance Light yellow to Yellow powder to crystal
Purity (GC) min. 98.0 %
Melting point 185.0 to 189.0 deg-C
Solubility in hot Methanol almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR