3-Carboxy-5-fluorophenylboronic acid ≥98%
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
H52886.03EA
3690
DKK
H52886.03
H52886.MD
3-Carboxy-5-fluorophenylboronic acid ≥98%
3-Carboxy-5-fluorophenylboronic acid
Formel:
C₇H₆BFO₄ Molekylvægt: 183,93 Smeltepunkt: 264…266 °C Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD07363791 CAS-nummer: 871329-84-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...