2-Carboxy-4-fluorobenzeneboronic acid, 97%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

H53180.MDEA 516 DKK
ALFAH53180.MD ALFAH53180.03
2-Carboxy-4-fluorobenzeneboronic acid, 97%
Purity: 97%
CAS No.: 874290-63-8
Molecular Formula: C7H6BFO4
Formula Weight: 183.93
Melting Point: 183-186°
MDL No.: MFCD08235078
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR