5-(2,3-Dimethylphenylcarbamoyl)-2-fluorobenzeneboronic acid, 97%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

H54692.03EA 3960 DKK
ALFAH54692.03 ALFAH54692.MD
5-(2,3-Dimethylphenylcarbamoyl)-2-fluorobenzeneboronic acid, 97%
Purity: 97%
Molecular Formula: C15H15BFNO3
Formula Weight: 287.09
MDL No.: MFCD20040235
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR