3-(2,3-Dimethylphenylcarbamoyl)-5-fluorobenzeneboronic acid, 97%
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
H54866.03EA 3960 DKK
ALFAH54866.03 ALFAH54866.MD
3-(2,3-Dimethylphenylcarbamoyl)-5-fluorobenzeneboronic acid, 97%
Purity: 97%
Molecular Formula: C15H15BFNO3
Formula Weight: 287.09
MDL No.: MFCD20040259