2-Chloro-4-(4-fluoro-3-nitrophenylcarbamoyl)benzeneboronic acid, 97%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

H55623.MDEA 988 DKK
ALFAH55623.MD ALFAH55623.03
2-Chloro-4-(4-fluoro-3-nitrophenylcarbamoyl)benzeneboronic acid, 97%
Purity: 97%
Molecular Formula: C13H9BClFN2O5
Formula Weight: 338.48
MDL No.: MFCD20040317
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR