2-Chloro-4-(4-nitrophenylcarbamoyl)benzeneboronic acid, 97%
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
H55341.03EA 3960 DKK
ALFAH55341.03 ALFAH55341.MD
2-Chloro-4-(4-nitrophenylcarbamoyl)benzeneboronic acid, 97%
Purity: 97%
Molecular Formula: C13H10BClN2O5
Formula Weight: 320.49
MDL No.: MFCD22125140