2-Chloro-4-(4-methyl-2-nitrophenylcarbamoyl)benzeneboronic acid, 97%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

H55323.03EA 4550 DKK
ALFAH55323.03 ALFAH55323.MD
2-Chloro-4-(4-methyl-2-nitrophenylcarbamoyl)benzeneboronic acid, 97%
Purity: 97%
Molecular Formula: C14H12BClN2O5
Formula Weight: 334.52
MDL No.: MFCD22125138
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR