Methyl propyl sulphide ≥98.0% (by GC)
Leverandør: TCI EUROPE
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Farlig
|
TCIAM2285-25GEA
501
DKK
TCIAM2285-25G
Methyl propyl sulphide ≥98.0% (by GC)
Methyl propyl sulphide
Formel:
C₄H₁₀S Kogepunkt: 97°C Smeltepunkt: -113°C Flammepunkt: 11°C |
MDL Number:
MFCD00015219 CAS-nummer: 3877-15-4 EINECS: 223-403-5 UN nummer: 1993 ADR: 3,II |
![]() |
Specifikationer Testresultater
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...