N,N,N',N'-Tetrakis(2-hydroxyethyl)ethylenediamine >50.0% (by GC)

Leverandør: TCI EUROPE
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

TCIAT0120-25GEA 189 DKK
TCIAT0120-25G
N,N,N',N'-Tetrakis(2-hydroxyethyl)ethylenediamine >50.0% (by GC)
N,N,N',N'-Tetrakis(2-hydroxyethyl)ethylenediamine
Formel: C₁₀H₂₄N₂O₄
Flammepunkt: 99°C
MDL Number: MFCD00002854
CAS-nummer: 140-07-8
EINECS: 205-396-0
https://dk.vwr.com/stibo/web/std.lang.all/44/48/28994448.jpg

Bestil nu

Specifikationer Testresultater

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 50.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR