2-Chloroethyl methyl sulphide ≥98.0% (by GC)
Leverandør: TCI EUROPE
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Farlig
|
TCIAC0588-10GEA
813
DKK
TCIAC0588-10G
2-Chloroethyl methyl sulphide ≥98.0% (by GC)
2-Chloroethyl methyl sulphide
Formel:
C₃H₇ClS Kogepunkt: 140°C Flammepunkt: 42°C Opbevaringstemperatur: Køleskab |
MDL Number:
MFCD00000978 CAS-nummer: 542-81-4 EINECS: 208-828-6 UN nummer: 2924 ADR: 3,III |
![]() |
Specifikationer Testresultater
Appearance | Colorless to Light yellow to Light orange clear liquid |
Purity (GC) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...