Acetic anhydride-[2,2'-¹³C₂] (99% ¹³C )
Leverandør: CAMBRIDGE ISOTOPE LABS., INC.
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
CAMBCLM-1160-0.25EA
0
DKK
CAMBCLM-1160-0.25
Acetic anhydride-[2,2'-¹³C₂] (99% ¹³C )
Acetic anhydride-[2,2'-¹³C₂]
Formel:
C₄H₆O₃ |
MDL Number:
MFCD00083863 CAS-nummer: 98006-45-2 UN nummer: 1715 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...