Cholesterol methyl carbonate ≥80.0% (by GC)
Leverandør: TCI EUROPE
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
TCIAC0715-10GEA
1110
DKK
TCIAC0715-10G
Cholesterol methyl carbonate ≥80.0% (by GC)
Cholesterol methyl carbonate
Formel:
C₂₉H₄₈O₃ Smeltepunkt: 114°C |
MDL Number:
MFCD00021136 CAS-nummer: 15507-52-5 |
![]() |
Specifikationer Testresultater
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 80.0 % |
Mesomorphic range | 100.0 to 110.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...