Furfuryl methyl sulphide ≥98.0% (by GC)
Leverandør: TCI EUROPE
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
TCIAF0245-25GEA
468
DKK
TCIAF0245-25G
Furfuryl methyl sulphide ≥98.0% (by GC)
Furfuryl methyl sulphide
Formel:
C₆H₈OS Molekylvægt: 128,19 g/mol Flammepunkt: 63°C |
MDL Number:
MFCD00009604 CAS-nummer: 1438-91-1 EINECS: 215-874-0 |
Specifikationer Testresultater
Purity (GC) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...