Furfuryl disulphide ≥95.0% (by GC)

Leverandør: TCI EUROPE
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

TCIAD1384-25GEA 632 DKK
TCIAD1384-25G
Furfuryl disulphide ≥95.0% (by GC)
Furfuryl disulphide
Formel: C₁₀H₁₀O₂S₂
Molekylvægt: 226,3152 g/mol
Kogepunkt: 121°C
Opbevaringstemperatur: Køleskab
MDL Number: MFCD00010082
CAS-nummer: 4437-20-1
EINECS: 224-649-6
https://dk.vwr.com/stibo/web/std.lang.all/82/34/28978234.jpg

Bestil nu

Specifikationer Testresultater

Appearance White to Yellow to Green clear liquid
Purity (GC) min. 95.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR