Methylmagnesium chloride 20% in THF til syntese, Sigma-Aldrich®
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Farlig
|
M-Clarity™ quality level = MQ200
8.20310.0250EA
1100
DKK
8.20310.0250
Methylmagnesium chloride 20% in THF til syntese, Sigma-Aldrich®
Methylmagnesium chloride
Formel:
CH₃ClMg Molekylvægt: 74,79 g/mol Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00000468 CAS-nummer: 676-58-4 EINECS: 211-629-7 UN nummer: 3399 ADR: 4.3 (3),I |
Specifikationer Testresultater
Solubility | (20 °C) (rigorous decomposition) |
Boiling Point | 65 - 66 °C (1013 hPa) (THF) |
Vapor Pressure | 200 hPa (20 °C) |
Flash Point | -20 °C (THF) |
Explosion Limit | 1.5 %(V) (THF) |
Density | 1.03 g/cm3 (20 °C) |
pH |
|
Ignition Point | 215 °C (THF) |
Synonyms | Chloromethylmagnesium |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...