Flame-retardant high visibility fleece jacket, Valier 9896
Leverandør: Sioen
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
113-7348EA 1510 DKK
SIOE9896/278/L SIOE9896/278/3XL SIOE9896/278/1XL SIOE9896/278/2XL SIOE9896/278/S SIOE9896/278/M
Flame-retardant high visibility fleece jacket, Valier 9896
This anti-static jacket is made of soft double-sided fleece (49,75% cotton, 49,75% Modacryl, 0,5% carbon) and can be zipped into different jackets. It provides high visibility protection against cold and flames.
  • Anti-pilling treatment, stitched seams
  • Straight collar, zip closure under flap with touch and close fastening
  • Two inset pockets with zip closure
  • Round inset sleeves, elasticated cuffs, elastic drawstring in hem
Back length: 77 cm
Weight: 300 g/m²
Certificeringer: EN 11612 (A1 B1 C2), EN 14116 (index 3), EN 1149-5, EN 14058 (1XXXX), EN 471 (2-2)