Cover spectacles, B-Line
Leverandør: Bollé Safety
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
B-Line
111-1586EA 253 DKK
111-1586 111-0343 111-0979
Cover spectacles, B-Line
Briller
Cover glasses (OTG) offering a panoramic field of vision.
  • Nylon frame
  • Scratch resistant
  • Wide side shields
Certificeringer: EN 166, (EN 170, EN 172)